1-Benzyl-2-amino-3-tert-butoxycarbonyl-4,5-dimethylpyrrole ≥95%

Supplier: Matrix Scientific

069211-1G 069211-5G 069211-25G
101939-284EA 349.23 USD
101939-284 101939-286 101939-288
1-Benzyl-2-amino-3-tert-butoxycarbonyl-4,5-dimethylpyrrole ≥95%
1-Benzyl-2-amino-3-tert-butoxycarbonyl-4,5-dimethylpyrrole

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₂
MDL Number: MFCD06660116
CAS Number: 100066-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR