Diethyl (1-trityl-1H-imidazol-4-yl)methylphosphonate ≥95%

Supplier: Matrix Scientific

069209-500MG 069209-1G 069209-5G
101939-272EA 756.92 USD
101939-272 101939-274 101939-276
Diethyl (1-trityl-1H-imidazol-4-yl)methylphosphonate ≥95%
Diethyl (1-trityl-1H-imidazol-4-yl)methylphosphonate

Hazard: Irritant

Formula: C₂₇H₂₉N₂O₃P
MDL Number: MFCD06660114
CAS Number: 473659-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR