3,3-Bis(fluoromethyl)oxetane ≥95%

Supplier: Matrix Scientific

069207-250MG 069207-500MG
101939-264EA 1309.23 USD
101939-264 101939-266
3,3-Bis(fluoromethyl)oxetane ≥95%
3,3-Bis(fluoromethyl)oxetane

Hazard: Irritant

Formula: C₅H₈F₂O
MDL Number: MFCD17170024
CAS Number: 338-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR