tert-Butyl-4-(aminomethyl)-4-fluoropiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
069204-250MG
069204-500MG
101939-256EA
29.23
USD
101939-256
101939-258
tert-Butyl-4-(aminomethyl)-4-fluoropiperidine-1-carboxylate ≥95%
tert-Butyl-4-(aminomethyl)-4-fluoropiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₁H₂₁FN₂O₂ |
MDL Number:
MFCD11847970 CAS Number: 620611-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...