Tetrahydro-2H-thiopyran-3-carboxylic acid ≥96%

Supplier: Matrix Scientific

069199-500MG 069199-1G
101939-244EA 1106.15 USD
101939-244 101939-246
Tetrahydro-2H-thiopyran-3-carboxylic acid ≥96%
Tetrahydro-2H-thiopyran-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₁₀O₂S
MDL Number: MFCD14584577
CAS Number: 161404-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR