(1-Methylcyclobutyl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

069190-250MG 069190-500MG 069190-1G
101939-238EA 756.92 USD
101939-238 101939-240 101939-242
(1-Methylcyclobutyl)methanamine hydrochloride ≥95%
(1-Methylcyclobutyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClN
MDL Number: MFCD18074449
CAS Number: 1245647-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR