Allylboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

069183-5G 069183-25G
101939-222EA 103.08 USD
101939-222 102833-454
Allylboronic acid pinacol ester ≥95%
Allylboronic acid pinacol ester

Hazard: Flammable/Moisture Sensitive/Stored under Argon/Store in Freezer

Formula: C₉H₁₇BO₂
Boiling Pt: 50…53 °C (5 mmHg)
Density: 0.89
Flash Pt: 46 °C
MDL Number: MFCD00013347
CAS Number: 72824-04-5
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR