(S)-N,N-Dimethyl(pyrrolidin-2-yl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

069178-1G 069178-5G
101939-212EA 1861.54 USD
101939-212 101939-214
(S)-N,N-Dimethyl(pyrrolidin-2-yl)methanamine hydrochloride ≥95%
(S)-N,N-Dimethyl(pyrrolidin-2-yl)methanamine hydrochloride

Hazard: Irritant

Formula: C₇H₁₇ClN₂
MDL Number: MFCD19443946
CAS Number: 1152113-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR