(3-Methylpyrrolidin-3-yl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

069176-1G 069176-5G
101939-204EA 2384.62 USD
101939-204 101939-206
(3-Methylpyrrolidin-3-yl)methanamine hydrochloride ≥95%
(3-Methylpyrrolidin-3-yl)methanamine hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂
MW: 150.65 g/mol
MDL Number: MFCD19443964
CAS Number: 1313738-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR