N,N-Dimethyl(1-methylpyrrolidin-3-yl)methanamine ≥95%

Supplier: Matrix Scientific

069174-1G 069174-5G 069174-25G
101939-198EA 2326.15 USD
101939-198 101939-200 101939-202
N,N-Dimethyl(1-methylpyrrolidin-3-yl)methanamine ≥95%
N,N-Dimethyl(1-methylpyrrolidin-3-yl)methanamine

Hazard: Irritant

Formula: C₈H₁₈N₂
MDL Number: MFCD19443943
CAS Number: 1313738-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR