N,N-Dimethylazetidin-3-amine hydrochloride ≥95%
Supplier: Matrix Scientific
069172-1G
069172-5G
101939-188EA
844.62
USD
101939-188
101939-190
N,N-Dimethylazetidin-3-amine hydrochloride ≥95%
N,N-Dimethylazetidin-3-amine hydrochloride
Hazard: Irritant
Formula:
C₅H₁₃ClN₂ |
MDL Number:
MFCD12405078 CAS Number: 935670-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...