2,4-Dichloropyrrolo[2,1-f][1,2,4]triazine ≥95%

Supplier: Matrix Scientific

069171-1G 069171-5G
101939-184EA 315.38 USD
101939-184 101939-186
2,4-Dichloropyrrolo[2,1-f][1,2,4]triazine ≥95%
2,4-Dichloropyrrolo[2,1-f][1,2,4]triazine

Hazard: Irritant

Formula: C₆H₃Cl₂N₃
MW: 188.0100 g/mol
MDL Number: MFCD11044885
CAS Number: 918538-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR