(S)-1-Boc-2-ethylpiperazine ≥98%

Supplier: Matrix Scientific

Synonyms: (2S)-2-Ethylpiperazine, N1-BOC protected, (S)-tert-Butyl 2-ethylpiperazine-1-carboxylate

069170-1G 069170-5G
101939-180EA 243.08 USD
101939-180 101939-182
(S)-1-Boc-2-ethylpiperazine ≥98%
(S)-1-Boc-2-ethylpiperazine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
MDL Number: MFCD07772100
CAS Number: 325145-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR