1-Boc-4-ethylaminopiperidine ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Boc-4-Ethylaminopiperidine, 4-(Ethylamino)piperidine, N1-BOC protected

069169-500MG 069169-1G 069169-5G
101939-174EA 640 USD
101939-174 101939-176 101939-178
1-Boc-4-ethylaminopiperidine ≥98%
1-Boc-4-ethylaminopiperidine

Hazard: Irritant

Formula: C₁₂H₂₄N₂O₂
MW: 228.34 g/mol
MDL Number: MFCD06808580
CAS Number: 264905-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR