7-Chlorothieno[3,2-b]pyridine ≥95%

Supplier: Matrix Scientific

069167-1G 069167-5G
101939-166EA 1338.46 USD
101939-166 101939-168
7-Chlorothieno[3,2-b]pyridine ≥95%
7-Chlorothieno[3,2-b]pyridine

Hazard: Irritant

Formula: C₇H₄ClNS
MDL Number: MFCD08448161
CAS Number: 69627-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR