Ethyl-6-bromopyrrolo[1,2-c]pyrimidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-bromopyrrolo[1,2-c]pyrimidine-3-carboxylate

069166-1G 069166-5G
101939-162EA 1426.15 USD
101939-162 101939-164
Ethyl-6-bromopyrrolo[1,2-c]pyrimidine-3-carboxylate ≥95%
Ethyl-6-bromopyrrolo[1,2-c]pyrimidine-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₉BrN₂O₂
MW: 269.1 g/mol
MDL Number: MFCD09878714
CAS Number: 588720-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR