6-Bromoquinolin-2(1H)-one ≥95%
Supplier: Matrix Scientific
069165-1G
069165-5G
101939-158EA
815.38
USD
101939-158
101939-160
6-Bromoquinolin-2(1H)-one ≥95%
Hazard: Irritant
Formula:
C₉H₆BrNO |
MDL Number:
MFCD08703169 CAS Number: 1810-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...