Ethyl-2-(5-bromobenzofuran-3-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(5-bromobenzofuran-3-yl)acetate

069163-1G 069163-5G
101939-150EA 1629.23 USD
101939-150 101939-152
Ethyl-2-(5-bromobenzofuran-3-yl)acetate ≥95%
Ethyl-2-(5-bromobenzofuran-3-yl)acetate

Hazard: Irritant

Formula: C₁₂H₁₁BrO₃
MW: 283.13 g/mol
MDL Number: MFCD09878709
CAS Number: 200204-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR