(R)-(-)-3-Amino-1-boc-piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-butyl (3R)-3-aminopiperidine-1-carboxylate, (R)-1-Boc-3-Aminopiperidine

069162-5G 069162-25G
101939-148EA 52.31 USD
101939-148 102663-842
(R)-(-)-3-Amino-1-boc-piperidine ≥95%
(R)-(-)-3-Amino-1-Boc-piperidine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD03094717
CAS Number: 188111-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR