4H-Amino-6-bromo-2,3-dihydrothiochromen-1,1-dioxide ≥95%

Supplier: Matrix Scientific

069161-1G 069161-5G
101939-142EA 1920 USD
101939-142 101939-144
4H-Amino-6-bromo-2,3-dihydrothiochromen-1,1-dioxide ≥95%
4H-Amino-6-bromo-2,3-dihydrothiochromen-1,1-dioxide

Hazard: Irritant

Formula: C₉H₁₀BrNO₂S
MDL Number: MFCD09025782
CAS Number: 916420-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR