1-Amino-1H-pyrrole-2-carbonitrile ≥95%

Supplier: Matrix Scientific

069160-1G 069160-5G 069160-25G
101939-136EA 756.92 USD
101939-136 101939-138 101939-140
1-Amino-1H-pyrrole-2-carbonitrile ≥95%
1-Amino-1H-pyrrole-2-carbonitrile

Hazard: Irritant

Formula: C₅H₅N₃
MDL Number: MFCD09025773
CAS Number: 159326-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR