2-Chloro-3-methoxypyrazine ≥95%
Supplier: Matrix Scientific
069158-5G
069158-25G
101939-128EA
250.77
USD
101939-128
102833-452
2-Chloro-3-methoxypyrazine ≥95%
2-Chloro-3-methoxypyrazine
Hazard: Irritant
Formula:
C₅H₅ClN₂O Melting Pt: 31…32 °C |
MDL Number:
MFCD02684324 CAS Number: 40155-28-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...