Ethyl-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5,6,7,8-Tetrahydro-1,2,4-triazolo[4,3-a]pyrazine-3-carboxylate

069157-1G 069157-5G
101939-122EA 1560 USD
101939-122 101939-124
Ethyl-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate ≥95%
Ethyl-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate

Hazard: Irritant

Formula: C₈H₁₂N₄O₂
MW: 196.2100 g/mol
MDL Number: MFCD08273912
CAS Number: 723286-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR