1-(1,3-Dimethyl-1H-pyrazol-5-yl)ethanone ≥98%
Supplier: Matrix Scientific
069156-1G
069156-5G
101939-118EA
640
USD
101939-118
101939-120
1-(1,3-Dimethyl-1H-pyrazol-5-yl)ethanone ≥98%
1-(1,3-Dimethyl-1H-pyrazol-5-yl)ethanone
Hazard: Irritant
Formula:
C₇H₁₀N₂O |
CAS Number:
87375-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...