4-Amino-3,5-dimethylpyridine ≥98%

Supplier: Matrix Scientific

069153-1G 069153-5G
101939-104EA 323.08 USD
101939-104 101939-106
4-Amino-3,5-dimethylpyridine ≥98%
4-Amino-3,5-dimethylpyridine

Hazard: Irritant

Formula: C₇H₁₀N₂
MDL Number: MFCD03413657
CAS Number: 43078-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR