3,6-Dibromopyridazine ≥98%

Supplier: Matrix Scientific

069149-5G 069149-25G
101939-094EA 281.54 USD
101939-094 101939-096
3,6-Dibromopyridazine ≥98%
3,6-Dibromopyridazine

Hazard: Irritant

Formula: C₄H₂Br₂N₂
MDL Number: MFCD00233947
CAS Number: 17973-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR