3-Hydroxy-5-trifluoromethylthiophene-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-5-(trifluoromethyl)thiophene-2-carboxylic acid

069147-1G 069147-5G
101939-082EA 640 USD
101939-082 101939-084
3-Hydroxy-5-trifluoromethylthiophene-2-carboxylic acid ≥97%
3-Hydroxy-5-trifluoromethylthiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₆H₃F₃O₃S
MW: 212.15 g/mol
MDL Number: MFCD05864365
CAS Number: 217959-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR