2,4-Dichloro-6-methylpyrimidin-5-ylamine ≥98%
Supplier: Matrix Scientific
Synonyms:
2,4-Dichloro-6-methylpyrimidin-5-amine, 5-Amino-2,4-dichloro-6-methylpyrimidine
069146-1G
069146-5G
101939-078EA
15.38
USD
101939-078
101939-080
2,4-Dichloro-6-methylpyrimidin-5-ylamine ≥98%
2,4-Dichloro-6-methylpyrimidin-5-ylamine
Hazard: Irritant
Formula:
C₅H₅Cl₂N₃ MW: 178.02 g/mol |
MDL Number:
MFCD08062586 CAS Number: 13162-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...