tert-Butyl-4-(7-hydroxypyrazolo[1,5-a]pyrimidin-2-yl)piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
069142-250MG
069142-500MG
101939-066EA
815.38
USD
101939-066
101939-068
tert-Butyl-4-(7-hydroxypyrazolo[1,5-a]pyrimidin-2-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(7-hydroxypyrazolo[1,5-a]pyrimidin-2-yl)piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₂N₄O₃ MW: 318.38 g/mol |
MDL Number:
MFCD16618564 CAS Number: 1228631-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...