1-(2-Phthalimidobutyryl)chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(1,3-Dioxoisoindolin-2-yl)butanoyl chloride

069141-1G 069141-5G
101939-062EA 233.85 USD
101939-062 101939-064
1-(2-Phthalimidobutyryl)chloride ≥97%
1-(2-Phthalimidobutyryl)chloride

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₃
MW: 251.67 g/mol
MDL Number: MFCD02656536
CAS Number: 10314-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR