1-(2-Phthalimidopropionyl)chloride ≥97%

Supplier: Matrix Scientific

069140-1G 069140-5G
101939-058EA 233.85 USD
101939-058 101939-060
1-(2-Phthalimidopropionyl)chloride ≥97%
1-(2-Phthalimidopropionyl)chloride

Hazard: Irritant

Formula: C₁₁H₈ClNO₃
MW: 237.64 g/mol
MDL Number: MFCD00023095
CAS Number: 17137-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR