2-(3-Piperidyl)acetonitrile hydrochloride ≥95%
Supplier: Matrix Scientific
069136-250MG
069136-1G
101939-050EA
1192.31
USD
101939-050
101939-052
2-(3-Piperidyl)acetonitrile hydrochloride ≥95%
2-(3-Piperidyl)acetonitrile hydrochloride
Hazard: Irritant
Formula:
C₇H₁₃ClN₂ |
MDL Number:
MFCD19443966 CAS Number: 5562-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...