tert-Butyl-4-(2-bromoacetyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(2-bromoacetyl)piperidine-1-carboxylate

069131-250MG 069131-1G
101939-040EA 52.31 USD
101939-040 101939-044
tert-Butyl-4-(2-bromoacetyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(2-bromoacetyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₀BrNO₃
MW: 306.21 g/mol
MDL Number: MFCD11975555
CAS Number: 301221-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR