tert-Butyl-11-oxo-5,10-diazaspiro[5.5]undecane-5-carboxylate ≥95%

Supplier: Matrix Scientific

069125-250MG 069125-500MG
101939-032EA 669.23 USD
101939-032 101939-034
tert-Butyl-11-oxo-5,10-diazaspiro[5.5]undecane-5-carboxylate ≥95%
tert-Butyl-11-oxo-5,10-diazaspiro[5.5]undecane-5-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₄N₂O₃
MW: 268.36 g/mol
MDL Number: MFCD16039652
CAS Number: 1221818-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR