4-(4-Bromophenyl)piperidine-4-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

069124-1G 069124-5G
101939-028EA 989.23 USD
101939-028 101939-030
4-(4-Bromophenyl)piperidine-4-carboxylic acid hydrochloride ≥95%
4-(4-Bromophenyl)piperidine-4-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅BrClNO₂
MDL Number: MFCD17015068
CAS Number: 1241725-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR