tert-Butyl-4-amino-4-(4-bromophenyl)piperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-amino-4-(4-bromophenyl)piperidine-1-carboxylate
069123-250MG
069123-1G
101939-024EA
1047.69
USD
101939-024
101939-026
tert-Butyl-4-amino-4-(4-bromophenyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-amino-4-(4-bromophenyl)piperidine-1-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₃BrN₂O₂ MW: 355.28 g/mol |
MDL Number:
MFCD11847939 CAS Number: 917925-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...