(S)-1-tert-Butyl-2-methyl-4-oxopiperidine-1,2-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-1-tert-Butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate

069120-250MG 069120-500MG 069120-1G
101939-010EA 872.31 USD
101939-010 101939-012 101939-014
(S)-1-tert-Butyl-2-methyl-4-oxopiperidine-1,2-dicarboxylate ≥95%
(S)-1-tert-Butyl-2-methyl-4-oxopiperidine-1,2-dicarboxylate

Hazard: Irritant

Formula: C₁₂H₁₉NO₅
MW: 257.29 g/mol
MDL Number: MFCD12547027
CAS Number: 756486-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR