tert-Butyl-4-pyrrolo[2,3-b]pyridin-1-ylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

069119-250MG 069119-500MG
101939-006EA 989.23 USD
101939-006 101939-008
tert-Butyl-4-pyrrolo[2,3-b]pyridin-1-ylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-pyrrolo[2,3-b]pyridin-1-ylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₃N₃O₂
MW: 301.39 g/mol
MDL Number: MFCD09839043
CAS Number: 1093759-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR