(5-Amino-1H-pyrazol-4-yl)(pyridin-2-yl)methanone ≥95%

Supplier: Matrix Scientific

069116-250MG 069116-500MG
101938-994EA 466.15 USD
101938-994 101938-996
(5-Amino-1H-pyrazol-4-yl)(pyridin-2-yl)methanone ≥95%
(5-Amino-1H-pyrazol-4-yl)(pyridin-2-yl)methanone

Hazard: Irritant

Formula: C₉H₈N₄O
MDL Number: MFCD12911775
CAS Number: 96219-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR