tert-Butyl-4-(5-chloro-1H-indazol-3-yl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(5-chloro-1H-indazol-3-yl)piperidine-1-carboxylate

069115-250MG 069115-500MG
101938-990EA 692.31 USD
101938-990 101938-992
tert-Butyl-4-(5-chloro-1H-indazol-3-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(5-chloro-1H-indazol-3-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₂ClN₃O₂
MW: 335.84 g/mol
MDL Number: MFCD12912623
CAS Number: 1198284-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR