tert-Butyl-4-(5-bromo-1H-indazol-3-yl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(5-bromo-1H-indazol-3-yl)piperidine-1-carboxylate

069112-250MG 069112-500MG
101938-978EA 960 USD
101938-978 101938-980
tert-Butyl-4-(5-bromo-1H-indazol-3-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(5-bromo-1H-indazol-3-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₂BrN₃O₂
MW: 380.29 g/mol
MDL Number: MFCD12912624
CAS Number: 1198284-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR