tert-Butyl-4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

069110-250MG 069110-500MG
101938-974EA 669.23 USD
101938-974 101938-976
tert-Butyl-4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₁ClFNO₃
MDL Number: MFCD16618544
CAS Number: 1228631-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR