1-tert-Butoxycarbonylindole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
069106-250MG
069106-500MG
101938-958EA
669.23
USD
101938-958
101938-960
1-tert-Butoxycarbonylindole-4-carboxylic acid ≥95%
1-tert-Butoxycarbonylindole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₅NO₄ |
MDL Number:
MFCD09834879 CAS Number: 848444-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...