3-Propoxyphenylboronic acid ≥98%

Supplier: Matrix Scientific

069099-1G 069099-5G
101938-928EA 263.08 USD
101938-928 101938-930
3-Propoxyphenylboronic acid ≥98%
3-Propoxyphenylboronic acid

Hazard: Irritant

Formula: C₉H₁₃BO₃
MDL Number: MFCD03427190
CAS Number: 149557-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR