1-Methyl-1,5,6,7-tetrahydroindazol-4-one ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-6,7-dihydro-1H-indazol-4(5H)-one

069093-1G 069093-5G
101938-900EA 815.38 USD
101938-900 101938-902
1-Methyl-1,5,6,7-tetrahydroindazol-4-one ≥97%
1-Methyl-1,5,6,7-tetrahydroindazol-4-one

Hazard: Irritant

Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
MDL Number: MFCD00930489
CAS Number: 85302-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR