5-Bromo-2-methylsulfanylpyrimidin-4-ol ≥96%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-(methylthio)pyrimidin-4(3H)-one

069091-1G 069091-5G
101938-890EA 230.77 USD
101938-890 101938-892
5-Bromo-2-methylsulfanylpyrimidin-4-ol ≥96%
5-Bromo-2-methylsulfanylpyrimidin-4-ol

Hazard: Irritant

Formula: C₅H₅BrN₂OS
MW: 221.08 g/mol
MDL Number: MFCD00223721
CAS Number: 81560-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR