3-Bromo-4,5-dihydroisoxazole-5-carboxylic acid amide ≥95%

Supplier: Matrix Scientific

069090-1G 069090-5G
101938-886EA 1512.31 USD
101938-886 101938-888
3-Bromo-4,5-dihydroisoxazole-5-carboxylic acid amide ≥95%
3-Bromo-4,5-dihydroisoxazole-5-carboxylic acid amide

Hazard: Irritant

Formula: C₄H₅BrN₂O₂
MDL Number: MFCD11707008
CAS Number: 1030613-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR