3-(Methylsulfonylamino)phenylboronic acid

Supplier: Matrix Scientific

069084-1G 069084-5G 069084-25G
101938-858EA 133.85 USD
101938-858 101938-860 101938-862
3-(Methylsulfonylamino)phenylboronic acid
3-(Methylsulfonylamino)phenylboronic acid

Hazard: Irritant

Formula: C₇H₁₀BNO₄S
Melting Pt: 90…96 °C
MDL Number: MFCD02179478
CAS Number: 148355-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR