2-(Trifluoromethyl)-5-pyridineboronic acid ≥96%

Supplier: Matrix Scientific

Synonyms: 6-(Trifluoromethyl)-3-pyridylboronic acid, 2-(Trifluoromethyl)pyridin-5-ylboronic acid, 2-(Trifluoromethyl)pyridine-5-boronic acid, (6-(Trifluoromethyl)pyridin-3-yl)boronic acid

069083-500MG 069083-1G
101938-852EA 53.85 USD
101938-852 101938-854
2-(Trifluoromethyl)-5-pyridineboronic acid ≥96%
2-(Trifluoromethyl)-5-pyridineboronic acid

Hazard: Irritant

Formula: C₆H₅BF₃NO₂
MW: 190.92 g/mol
MDL Number: MFCD07375381
CAS Number: 868662-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR