3-Iodo-1H-pyrrolo[2,3-c]pyridine ≥98%

Supplier: Matrix Scientific

069079-500MG 069079-1G
101938-836EA 1106.15 USD
101938-836 101938-838
3-Iodo-1H-pyrrolo[2,3-c]pyridine ≥98%
3-Iodo-1H-pyrrolo[2,3-c]pyridine

Hazard: Irritant

Formula: C₇H₅IN₂
MDL Number: MFCD10699183
CAS Number: 956003-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR